siRNA Details
Gene Symbol: Ccdc110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033246.2 | NP_001028418.1 | 5, 6 | 448 | ||
GenBank mRNA | AK132742.1 | - | 5, 6 | 449 | ||
AK161248.1 | - | 5, 6 | 436 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ccdc110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033246.2 | NP_001028418.1 | 5, 6 | 448 | ||
GenBank mRNA | AK132742.1 | - | 5, 6 | 449 | ||
AK161248.1 | - | 5, 6 | 436 |