siRNA Details
Gene Symbol: Atxn7l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098836.1 | NP_001092306.1 | 13 | 2461 | ||
NM_001098837.1 | NP_001092307.1 | 13 | 2440 | |||
GenBank mRNA | AK053165.1 | - | 1 | 548 | ||
BC059880.1 | - | 7 | 1856 | |||
BC130269.1 | - | 12 | 2212 |
