siRNA Details
Gene Symbol: Rnf157 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027258.1 | NP_081534.1 | 12 | 1543 | ||
GenBank mRNA | AK038042.1 | - | 6 | 770 | ||
AK122571.1 | - | 2 | 279 | |||
AK169566.1 | - | 7 | 723 | |||
AK182359.1 | - | 1 | 44 | |||
BC053070.1 | - | 12 | 1322 |
