siRNA Details
Gene Symbol: Fam208a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114879.1 | NP_001108351.1 | 24 | 6456 | ||
NM_028945.1 | NP_083221.1 | 24 | 6730 | |||
GenBank mRNA | AK173099.1 | - | 4 | 2192 | ||
BC064458.1 | - | 6 | 2792 |
