siRNA Details
Gene Symbol: 5031439G07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033273.2 | NP_001028445.1 | 10 | 4388 | ||
GenBank mRNA | AK030332.1 | - | 2143 | |||
AK149782.1 | - | 10 | 4389 | |||
AK160765.1 | - | 1 | 226 |
