siRNA Details
Gene Symbol: Brd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033274.3 | NP_001028446.2 | 6 | 2102 | ||
GenBank mRNA | AK149714.1 | - | 6 | 2114 | ||
AK220233.1 | - | 5 | 1023 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001108103, XM_006242194, XM_006242196, XM_006242197.
Gene Ontology Categories:
