siRNA Details
Gene Symbol: Myh6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017239.2 | NP_058935.2 | 2 | 219 | ||
GenBank mRNA | X15938.1 | - | 2 | 216 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Myh6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017239.2 | NP_058935.2 | 2 | 219 | ||
GenBank mRNA | X15938.1 | - | 2 | 216 |