siRNA Details
Gene Symbol: Alpk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037294.1 | NP_001032371.1 | 9 | 6094 | ||
GenBank mRNA | AK132555.1 | - | 6 | 4422 | ||
AY044451.1 | - | 5 | 3816 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Alpk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037294.1 | NP_001032371.1 | 9 | 6094 | ||
GenBank mRNA | AK132555.1 | - | 6 | 4422 | ||
AY044451.1 | - | 5 | 3816 |