siRNA Details
Gene Symbol: Oosp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033283.2 | NP_001028455.2 | 3 | 343 | ||
GenBank mRNA | AK143410.1 | - | 3 | 344 | ||
BC094934.1 | - | 3 | 305 | |||
BC141228.1 | - | 3 | 320 | 9055840 | ||
BC145553.1 | - | 2 | 141 |
