siRNA Details
Gene Symbol: BC026585 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033284.1 | NP_001028456.1 | 2, 3 | 180 | ||
GenBank mRNA | AK143913.1 | - | 2, 3 | 180 | ||
AK151310.1 | - | 2, 3 | 188 | |||
BC026585.1 | - | 2, 3 | 182 | |||
BC045198.1 | - | 2, 3 | 177 | 5065108 |
