siRNA Details
Gene Symbol: BC026585 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033284.1 | NP_001028456.1 | 10 | 1216 | ||
GenBank mRNA | AK143913.1 | - | 10 | 1216 | ||
AK151310.1 | - | 7 | 731 | |||
BC026585.1 | - | 10 | 1218 | |||
BC045198.1 | - | 10 | 1213 | 5065108 | ||
BC057972.1 | - | 4 | 718 |
