siRNA Details
Gene Symbol: Gm4846 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164306.1 | NP_001157778.1 | 5 | 821 | ||
GenBank mRNA | BC147800.1 | - | 5 | 821 | 9087517 | |
BC147804.1 | - | 5 | 829 | 9087521 |
