siRNA Details
Gene Symbol: Ppig | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081086.1 | NP_001074555.1 | 14 | 5140 | ||
GenBank mRNA | AK038755.1 | - | 1 | 758 | ||
AK045812.1 | - | 1 | 2395 | |||
AK049547.1 | - | 1 | 1328 | |||
AK164670.1 | - | 1 | 3135 |
