siRNA Details
Gene Symbol: Tnks1bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081260.2 | NP_001074729.1 | 6 | 3083 | ||
GenBank mRNA | AK129436.1 | - | 2 | 1776 | ||
BC025943.1 | - | 1 | 532 | |||
BC058639.1 | - | 2 | 2135 | |||
BC150947.1 | - | 6 | 3003 | 9087858 |
