siRNA Details
Gene Symbol: Hsd3b6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017265.4 | NP_058961.4 | 4 | 1267 | ||
GenBank mRNA | BC089937.1 | - | 4 | 1267 | 7309637 | |
L17138.1 | - | 3 | 1105 |
Gene Symbol: Hsd3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042619.1 | NP_001036084.1 | 4 | 1201 | ||
GenBank mRNA | M38179.1 | - | 4 | 1201 |
Gene Symbol: Hsd3b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007719.3 | NP_001007720.3 | 6 | 1201 | ||
GenBank mRNA | BC086578.1 | - | 6 | 1201 | 7306702 | |
DQ515797.1 | - | 1 | 373 | |||
M38178.1 | - | 5 | 1220 |
Gene Symbol: LOC100911154 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749361.2 | XP_003749409.1 | 5 | 1286 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001111336, NM_001161743, NM_013821, NM_001161742, NM_001161744, and others.
Gene Ontology Categories:
