siRNA Details
Gene Symbol: Ciart | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033302.2 | NP_001028474.1 | 6 | 1154 | ||
GenBank mRNA | AK164001.1 | - | 6 | 1154 | ||
BC132471.1 | - | 5 | 1050 | 40130748 | ||
BC138510.1 | - | 5 | 1050 | 8861530 |
