siRNA Details
Gene Symbol: Sh2d5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099631.1 | NP_001093101.1 | 10 | 2356 | ||
GenBank mRNA | AK032598.1 | - | 10 | 2288 | ||
DQ832275.1 | - | 10 | 2356 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sh2d5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099631.1 | NP_001093101.1 | 10 | 2356 | ||
GenBank mRNA | AK032598.1 | - | 10 | 2288 | ||
DQ832275.1 | - | 10 | 2356 |