siRNA Details
Gene Symbol: Ffar3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033316.2 | NP_001028488.1 | 2 | 974 | ||
GenBank mRNA | AK148616.1 | - | 2 | 975 | ||
BC109330.2 | - | 1 | 1187 | 40051470 | ||
BC125009.1 | - | 1 | 1187 | 40051462 |
