siRNA Details
Gene Symbol: BC030336 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164580.1 | NP_001158052.1 | 3 | 1913 | ||
GenBank mRNA | AK034119.1 | - | 1 | 38 | ||
AK148518.1 | - | 1 | 1210 | |||
AK148546.1 | - | 1 | 1240 | |||
AK164547.1 | - | 1 | 1250 |
