siRNA Details
Gene Symbol: BC030336 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164580.1 | NP_001158052.1 | 3 | 1761 | ||
GenBank mRNA | AK148518.1 | - | 1 | 1058 | ||
AK148546.1 | - | 1 | 1088 | |||
AK164547.1 | - | 1 | 1098 |
