siRNA Details
Gene Symbol: Slc25a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017307.3 | NP_059003.1 | 3, 4 | 424 | ||
GenBank mRNA | BC100077.1 | - | 3, 4 | 408 | 7126394 |

More Information
Other Species Targeted

Gene Ontology Categories:
