siRNA Details
Gene Symbol: 4930596D02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033766.3 | NP_001028938.1 | 2 | 214 | ||
GenBank mRNA | AK133137.1 | - | 2 | 215 | ||
BC147602.1 | - | 2 | 214 | 9056592 | ||
BC147613.1 | - | 2 | 214 | 9056603 |
