siRNA Details
Gene Symbol: Tspyl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085421.1 | NP_001078890.1 | 1 | 3690 | ||
GenBank mRNA | AK031988.1 | - | 1 | 3400 | ||
AK087510.1 | - | 1 | 3628 | |||
AK173253.1 | - | 1 | 3630 | |||
BC079547.1 | - | 1 | 3641 |
