siRNA Details
Gene Symbol: Ttc38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033337.4 | NP_001028509.2 | 9 | 902 | ||
GenBank mRNA | AK078848.1 | - | 9 | 863 | ||
AK133636.1 | - | 8 | 610 | |||
AK200042.1 | - | 3 | 154 | |||
AK204138.1 | - | 3 | 154 | |||
BC024550.1 | - | 9 | 852 | |||
BC030849.1 | - | 9 | 865 | |||
BC132621.1 | - | 9 | 887 | 40130898 | ||
BC132623.1 | - | 9 | 887 | 40130900 |
