siRNA Details
Gene Symbol: Arid1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085355.1 | NP_001078824.1 | 20 | 11066 | ||
GenBank mRNA | AK049919.1 | - | 1 | 3477 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Arid1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085355.1 | NP_001078824.1 | 20 | 11066 | ||
GenBank mRNA | AK049919.1 | - | 1 | 3477 |