siRNA Details
Gene Symbol: Gnao1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017327.1 | NP_059023.1 | 3, 4 | 1013 | ||
XM_008772320.1 | XP_008770542.1 | 3, 4 | 1022 | |||
XM_008772321.1 | XP_008770543.1 | 3, 4 | 1022 | |||
XM_008772322.1 | XP_008770544.1 | 2, 3 | 3631 | |||
GenBank mRNA | M12671.1 | - | 2, 3 | 177 | ||
M17526.1 | - | 3, 4 | 1013 |
