siRNA Details
Gene Symbol: Lrrc30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033340.3 | NP_001028512.2 | 1 | 1358 | ||
GenBank mRNA | AK137598.1 | - | 1 | 1359 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lrrc30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033340.3 | NP_001028512.2 | 1 | 1358 | ||
GenBank mRNA | AK137598.1 | - | 1 | 1359 |