siRNA Details
Gene Symbol: F830016B08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101475.2 | NP_001094945.1 | 2 | 1508 | ||
GenBank mRNA | AK156308.1 | - | 2 | 1514 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: F830016B08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101475.2 | NP_001094945.1 | 2 | 1508 | ||
GenBank mRNA | AK156308.1 | - | 2 | 1514 |