siRNA Details
Gene Symbol: Gpr161 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081126.1 | NP_001074595.1 | 4 | 928 | ||
GenBank mRNA | AK145635.1 | - | 3 | 953 | ||
AY255596.1 | - | 2 | 419 | |||
BC137659.1 | - | 3 | 866 | 8860678 | ||
EF197953.1 | - | 3 | 953 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
