siRNA Details
Gene Symbol: Prps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017243.2 | NP_058939.1 | 2 | 275 | ||
GenBank mRNA | BC078853.1 | - | 2 | 273 | 7190069 | |
CB702494.1 | - | 2 | 275 | |||
M17258.1 | - | 2 | 251 | |||
M29392.1 | - | 2 | 251 | |||
X16554.1 | - | 2 | 272 |
Gene Symbol: LOC314140 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009694.1 | NP_001009694.1 | 2 | 547 | ||
XM_008764657.1 | XP_008762879.1 | 1 | 323 | |||
GenBank mRNA | BC088149.1 | - | 2 | 547 | 7380332 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001037746, AK134500, BC110357.
Gene Ontology Categories:
