siRNA Details
Gene Symbol: 1700024P16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162980.1 | NP_001156452.1 | 20 | 2623 | ||
NM_001285952.1 | NP_001272881.1 | 21 | 2701 | |||
NM_001285953.1 | NP_001272882.1 | 20 | 2556 | |||
GenBank mRNA | AK028077.1 | - | 20 | 2563 | ||
AK133762.1 | - | 20 | 2495 | |||
AV174094.2 | - | 1 | 163 | |||
BC137723.1 | - | 20 | 2623 | 8860742 | ||
BC144857.1 | - | 21 | 2700 | 9053405 | ||
BC144858.1 | - | 21 | 2664 | 9053406 |
