siRNA Details
Gene Symbol: 1700024P16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162980.1 | NP_001156452.1 | 20 | 2904 | ||
NM_001285952.1 | NP_001272881.1 | 21 | 2982 | |||
NM_001285953.1 | NP_001272882.1 | 20 | 2837 | |||
GenBank mRNA | AK028077.1 | - | 20 | 2844 | ||
AK133762.1 | - | 20 | 2776 | |||
AV174094.2 | - | 1 | 444 |
