siRNA Details
Gene Symbol: Rab42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081651.1 | NP_001075120.1 | 2 | 353 | ||
GenBank mRNA | AB232642.1 | - | 2 | 334 | ||
AK035722.1 | - | 1 | 698 | |||
BC132060.1 | - | 2 | 382 | 40130337 | ||
BC132062.1 | - | 2 | 382 | 40130339 |
