siRNA Details
Gene Symbol: Ptchd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083342.1 | NP_001076811.1 | 21 | 7101 | ||
GenBank mRNA | AK160846.1 | - | 1 | 2082 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ptchd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083342.1 | NP_001076811.1 | 21 | 7101 | ||
GenBank mRNA | AK160846.1 | - | 1 | 2082 |