siRNA Details
Gene Symbol: Ptchd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083342.1 | NP_001076811.1 | 21 | 4861 | ||
GenBank mRNA | AK078196.1 | - | 11 | 2359 | ||
AK090326.1 | - | 21 | 4887 | |||
AK173152.1 | - | 20 | 4304 | |||
BC058128.1 | - | 2 | 1031 | |||
BC070441.1 | - | 21 | 4735 | 30535318 | ||
BC072569.1 | - | 21 | 4760 | 5719275 |
