siRNA Details
Gene Symbol: 2010107G12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025573.2 | NP_001020744.2 | 8 | 1969 | ||
GenBank mRNA | AK008337.1 | - | 1 | 240 | ||
BC075651.1 | - | 8 | 1969 | 30541436 |
