siRNA Details
Gene Symbol: Cyp2b23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081148.1 | NP_001074617.1 | 9 | 2122 | ||
GenBank mRNA | BC100376.1 | - | 4 | 1250 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cyp2b23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081148.1 | NP_001074617.1 | 9 | 2122 | ||
GenBank mRNA | BC100376.1 | - | 4 | 1250 |