siRNA Details
Gene Symbol: Zfp568 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033355.3 | NP_001028527.2 | 10 | 4884 | ||
NM_001167872.1 | NP_001161344.1 | 10 | 4891 | |||
NM_001167873.1 | NP_001161345.1 | 10 | 4888 | |||
GenBank mRNA | BC082606.1 | - | 9 | 4575 | 30672810 |
