siRNA Details
Gene Symbol: Atr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019864.1 | NP_063917.1 | 5 | 1341 | ||
GenBank mRNA | AK139834.1 | - | 5 | 1319 | ||
AK142244.1 | - | 5 | 1301 | |||
AK157754.1 | - | 5 | 1289 | |||
AK157757.1 | - | 5 | 1341 | |||
AK170039.1 | - | 4 | 2009 | |||
AK171072.1 | - | 5 | 1273 |
