siRNA Details
Gene Symbol: LOC100911116 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749360.3 | XP_003749408.1 | 4 | 1248 |
Gene Symbol: Hsd3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042619.1 | NP_001036084.1 | 4 | 1254 | ||
GenBank mRNA | M38179.1 | - | 4 | 1254 |
Gene Symbol: Hsd3b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007719.3 | NP_001007720.3 | 6 | 1254 | ||
GenBank mRNA | BC086578.1 | - | 6 | 1254 | 7306702 | |
M38178.1 | - | 5 | 1273 |
Gene Symbol: Hsd3b5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012584.1 | NP_036716.1 | 1186 | |||
GenBank mRNA | BC092571.1 | - | 1203 | 7369915 | ||
M67465.1 | - | 1186 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
