siRNA Details
Gene Symbol: Psma2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017279.1 | NP_058975.1 | 5 | 437 | ||
GenBank mRNA | BC060576.1 | - | 5 | 456 | 6919255 | |
FQ214776.1 | - | 5 | 452 | |||
FQ223872.1 | - | 452 | ||||
J02897.1 | - | 5 | 437 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008944, BC026768, CJ052982, X70303, AK012119, and others.
Gene Ontology Categories:
