siRNA Details
Gene Symbol: Mroh5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033365.2 | NP_001028537.1 | 11 | 1676 | ||
GenBank mRNA | AK158799.1 | - | 11 | 1677 | ||
BC150895.1 | - | 10 | 1579 | 9087806 | ||
BC172046.1 | - | 10 | 1579 | 9054332 |
