siRNA Details
Gene Symbol: Cntrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012018.2 | NP_036148.2 | 33 | 5530 | ||
GenBank mRNA | AK131139.1 | - | 11 | 1884 | ||
BC023035.1 | - | 4 | 789 | |||
BC079536.1 | - | 21 | 3555 | |||
BC158010.1 | - | 32 | 5403 | 9007318 |
