siRNA Details
Gene Symbol: Gm648 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033372.2 | NP_001028544.1 | 3, 4 | 674 | ||
GenBank mRNA | AK146653.1 | - | 3, 4 | 675 | ||
BC147598.1 | - | 3, 4 | 674 | 9056588 | ||
BC147617.1 | - | 3, 4 | 674 | 9056607 |
