siRNA Details
Gene Symbol: Zfp616 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177570.1 | NP_001171041.1 | 4 | 2123 | ||
GenBank mRNA | BC083001.1 | - | 4 | 2125 |
Gene Symbol: Gm5665 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK139826.1 | - | 1235 |
