siRNA Details
Gene Symbol: Gm9975 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK050463.1 | - | 1 | 1387 | ||
BC042188.1 | - | 1 | 231 | |||
BC096608.1 | - | 1 | 1716 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm9975 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK050463.1 | - | 1 | 1387 | ||
BC042188.1 | - | 1 | 231 | |||
BC096608.1 | - | 1 | 1716 |