siRNA Details
Gene Symbol: 9330182O14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256056.1 | NP_001242985.1 | 4 | 1370 | ||
GenBank mRNA | AK034361.1 | - | 4 | 1372 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 9330182O14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256056.1 | NP_001242985.1 | 4 | 1370 | ||
GenBank mRNA | AK034361.1 | - | 4 | 1372 |