siRNA Details
Gene Symbol: Bsph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033418.4 | NP_001028590.1 | 5 | 560 | ||
NM_001301682.1 | NP_001288611.1 | 4 | 530 | |||
GenBank mRNA | AK136662.1 | - | 5 | 561 | ||
BC147454.1 | - | 5 | 545 | 9056096 | ||
BC147459.1 | - | 5 | 545 | 9056101 | ||
BC171987.1 | - | 4 | 515 | 9054273 | ||
BC171991.1 | - | 4 | 515 | 9054277 | ||
DQ227498.1 | - | 5 | 540 |
