siRNA Details
Gene Symbol: Fnd3c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033424.3 | NP_001028596.2 | 20 | 3509 | ||
GenBank mRNA | AK030291.1 | - | 20 | 3513 | ||
AK146031.1 | - | 20 | 3510 | |||
BC038164.1 | - | 1 | 580 |
