siRNA Details
Gene Symbol: Ksr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114545.2 | NP_001108017.2 | 4 | 1512 | ||
GenBank mRNA | AK137433.1 | - | 4 | 1514 | ||
BC068233.1 | - | 2 | 297 | |||
BC150883.1 | - | 4 | 1512 | |||
BC150890.1 | - | 4 | 1512 | |||
DQ531035.1 | - | 4 | 685 |
